Order Entry
Czech Republic
ContactUsLinkComponent
4,4-Difluoro-L-prolinamide hydrochloride 98%
Irritant
4,4-Difluoro-L-prolinamide hydrochloride 98%
Catalog # H27770.03
CAS Number:  
Irritant
4,4-Difluoro-L-prolinamide hydrochloride 98%
Catalog # H27770.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:426844-51-1
 :
Formula:C₅H₈F₂N₂O·HCl
MDL Number:MFCD06797014
 :186.59 g/mol
Melting Pt:221…223 °C (dec.)
 :GHS07
Storage Temperature:Ambient