
Triethylsulphonium bis(trifluoromethanesulphonyl)imide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:321746-49-0
:
Formula:C₈H₁₅F₆NO₄S₃
MDL Number:MFCD08276438
:399.38 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together