
1-Methyl-1H-indazole-3-carboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50890-83-0
:
Formula:C₉H₈N₂O₂
MDL Number:MFCD00272569
:176.17 g/mol
Melting Pt:∼ 218 °C (dec.)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together