Order Entry
Czech Republic
ContactUsLinkComponent
cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene
Irritant
cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene
Catalog # H26815.03
CAS Number:  
Irritant
cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene
Catalog # H26815.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:79236-92-3
 :
Formula:C₁₀H₁₈N₄
MDL Number:MFCD09263319
 :194.28 g/mol
Melting Pt:29…31 °C
 :GHS07
Storage Temperature:Ambient