

4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid methyl ester 97%
Catalog # H26353.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:175202-48-9
:
Formula:C₈H₇NO₂S₂
MDL Number:MFCD00173757
:213.28 g/mol
Melting Pt:117…119 °C
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient