
4-Nitro-1H-pyrazole-3-carboxylic acid 98%
Catalog # H26224.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
Specifications
About this item
CAS Number:5334-40-7
:
Formula:C₄H₃N₃O₄
MDL Number:MFCD00090899
:157.085 g/mol
Melting Pt:∼ 195 °C (dec.)
:GHS07
Storage Temperature:Ambient