
Cyclopropane-1,1-dicarboxylic acid mono methyl ester 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:113020-21-6
:
Formula:C₆H₈O₄
MDL Number:MFCD06800496
:144.13 g/mol
Boiling Pt:85…87 °C (0.2 mmHg)
Melting Pt:50…57 °C
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together