Ethyl-4,4-difluorocyclohexanecarboxylate 97%
5 g
Supplier: Thermo Fisher Scientific
Ethyl-4,4-difluorocyclohexanecarboxylate 97%
Catalog # H25754.06
Supplier: Thermo Fisher Scientific
CAS Number: 178312-47-5
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 178312-47-5 Molecular Formula: C9H14F2O2 Formula Weight: 192.21 Boiling Point: 84-86°/8mm MDL No.: MFCD03788494
Synonyms: Ethyl 4,4-difluorocyclohexanecarboxylate
- CAS Number:178312-47-5
- Formula:C₉H₁₄F₂O₂
- MDL Number:MFCD03788494
- Molecular Weight:192.21 g/mol
- Boiling Pt:84…86 °C (8 mmHg)
- Storage Temperature:Ambient
Documents
Specifications
Specifications
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



