Methyl-1-cyclohexene-1-carboxylate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18448-47-0
:
Formula:C₈H₁₂O₂
MDL Number:MFCD00001544
:140.182 g/mol
Boiling Pt:190…192 °C
Density:1.030
Flash Pt:73 °C (165 °F)
Storage Temperature:Ambient
EINECS:242-331-5
Related Information
Frequently Bought Together