
Sodium phosphonoformate hexahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34156-56-4
:
Formula:Na₂O₃PCO₂Na·6H₂O
MDL Number:MFCD00150176
:300.08 g/mol (191.98 anhy)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together