

Ammonium cobalt(II) phosphate, anhydrous 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14590-13-7
:
Formula:H₄CoNO₄P
MDL Number:MFCD02683100
:171.94 g/mol
Merck Index:14,513
EINECS:238-633-1
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together