
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-08-7
:
Formula:C₂₁H₂₄O₄
MDL Number:MFCD00021235
:340.42 g/mol
Melting Pt:>300 °C
EINECS:201-003-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together