
1,2-Bis(tosyloxy)ethane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6315-52-2
:
Formula:C₁₆H₁₈O₆S₂
MDL Number:MFCD00008549
:370.43 g/mol
Melting Pt:122…127 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together