ContactUsLinkComponent
Perfluoro-1,3,5-trimethylcyclohexane mixture of isomers, tech.
Perfluoro-1,3,5-trimethylcyclohexane mixture of isomers, tech.
  B21500.18
 :  
Perfluoro-1,3,5-trimethylcyclohexane mixture of isomers, tech.
  B21500.18
 :  
  • Pk:
    50 g
  • Seq:
    3368
 :374-76-5
 :
 :C₉F₁₈
 :MFCD00066614
 :450.07 g/mol
 :125…128 °C
 :–68 °C
 :1.888
 :Ambient