Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-4,5-dibromo-3-hydroxythiophene-2-carboxylate 97%
Irritant
Methyl-4,5-dibromo-3-hydroxythiophene-2-carboxylate 97%
Catalog # B20780.06
CAS Number:  
Irritant
Methyl-4,5-dibromo-3-hydroxythiophene-2-carboxylate 97%
Catalog # B20780.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Melting Pt:125…128 °C
 :316.89 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:96232-71-2
Formula:C₆H₄Br₂O₃S
MDL Number:MFCD00052082