
4-Methyl-1,2,3-thiadiazole-5-carboxylic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18212-21-0
:
Formula:C₄H₄N₂O₂S
MDL Number:MFCD00173830
Melting Pt:184…186 °C
:144.15 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together