cis-3-Methylcyclohexanol 95%
Catalog # B20288.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:114.19 g/mol
Flash Pt:73 °C (163 °F)
Storage Temperature:Ambient
CAS Number:24965-90-0
Formula:C₇H₁₄O
MDL Number:MFCD00067155