
Isoquinoline-3-carboxylic acid monohydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207399-25-5
:203626-75-9
Formula:C₁₀H₉NO₃
MDL Number:MFCD00075137
:191.183 g/mol
Melting Pt:166…168 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together