
Phosphonitrilic chloride trimer 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:940-71-6
:
Formula:Cl₆N₃P₃
MDL Number:MFCD00006474
:347.64 g/mol
Boiling Pt:127 °C (13 mmHg)
Melting Pt:111…115 °C
Density:1.98
Storage Temperature:Ambient
EINECS:213-376-8
ADR:8,II
:GHS05
Related Information
Frequently Bought Together