7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2503-56-2
:
Formula:C₆H₆N₄O
MDL Number:MFCD00005555
:150.14 g/mol
Melting Pt:281…285 °C
Storage Temperature:Ambient
EINECS:219-706-7
Related Information
Frequently Bought Together