
Methyl-4-hydroxy-3,5-dimethoxybenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:884-35-5
:
Formula:C₁₀H₁₂O₅
MDL Number:MFCD00017199
:212.20 g/mol
Melting Pt:102…108 °C
EINECS:429-050-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together