12-Hydroxystearic acid ≥85%, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-14-9
:
Formula:C₁₈H₃₆O₃
MDL Number:MFCD00004592
:300.48 g/mol
Melting Pt:72…77 °C
Storage Temperature:Ambient
EINECS:203-366-1
Related Information
Frequently Bought Together