Solvent Blue 38
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1328-51-4
:
Formula:C32H12CuN8Na2O6S2
MDL Number:MFCD00071424
Melting Pt:∼ 235 °C (dec.)
:778.15 g/mol
Storage Temperature:Ambient
EINECS:215-523-1
Related Information
Frequently Bought Together