

Crotonoyl chloride 90% remainder mainly cis-isomer, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:625-35-4
Formula:C₄H₅ClO
MDL Number:MFCD00000734
:104.53 g/mol
Boiling Pt:121…123 °C
Density:1.091
Flash Pt:35 °C (95 °F)
Storage Temperature:Refrigerator
EINECS:210-889-9
UN:2920
ADR:8,II
:GHS05
:GHS02
Related Information
Frequently Bought Together