Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tris[4-(trifluoromethyl)phenyl]phosphine
Tris[4-(trifluoromethyl)phenyl]phosphine 98%
Tris[4-(trifluoromethyl)phenyl]phosphine 98%
A12885.03
:
Thermo Fisher Scientific
:
Tris[4-(trifluoromethyl)phenyl]phosphine 98%
A12885.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
13406-29-6
:
:
C₂₁H₁₂F₉P
:
MFCD00058883
:
466.28 g/mol
:
72…75 °C
:
GHS07
:
Ambient