

S,S'-Dimethyl-N-cyanodithioiminocarbonate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10191-60-3
:
Formula:C₄H₆N₂S₂
MDL Number:MFCD00009825
:146.23 g/mol
Melting Pt:50…56 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:233-467-6
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together