
Atropine sulphate monohydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5908-99-6
:
Formula:C₁₇H₂₃NO₃
MDL Number:MFCD00074815
:676.82 g/mol
Merck Index:14,875
Melting Pt:187…192 °C
Storage Temperature:Ambient
EINECS:200-235-0
ADR:6.1,II
:GHS06
Related Information
Frequently Bought Together