



Sodium cyclopentadienide 2-3 M in THF
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4984-82-1
:
Formula:C₅H₅Na
MDL Number:MFCD00012161
:88.085 g/mol
Density:0.938
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
EINECS:225-636-8
UN:3399
ADR:4.3,II
:GHS02
:GHS05
:GHS08
:GHS07
Related Information
Frequently Bought Together