


Sodium borohydride 4.4 M in aqueous solution stable
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16940-66-2
:
Formula:NaBH₄
MDL Number:MFCD00003518
:37.83 g/mol
Density:1.4
Storage Temperature:Ambient
EINECS:241-004-4
UN:3320
ADR:8,II
:GHS02
:GHS05
:GHS07
Related Information
Frequently Bought Together