Ruthenium ≥99.95% (metals basis), pellets 3-6 mm
Catalog # 55236.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
Specifications
About this item
:
:101.07 g/mol
Storage Temperature:Ambient
EINECS:231-127-1
CAS Number:7440-18-8
Formula:Ru
MDL Number:MFCD00011207