Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Dichloro bis(dicyclohexylphosphino)propane palladium(II) (17.3% Pt)
Catalog # 46780.03
CAS Number:  
Irritant
Dichloro bis(dicyclohexylphosphino)propane palladium(II) (17.3% Pt)
Catalog # 46780.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

CAS Number:1041005-52-0
 :
Formula:C₂₇H₅₀Cl₂P₂Pd
 :613.965 g/mol
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD14155689