
Dibromo(1,5-cyclooctadiene)palladium(II) (28.4% Pd)
Catalog # 46492.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
:
:374.412 g/mol
Melting Pt:201…205 °C
:GHS07
Storage Temperature:Ambient
CAS Number:12145-47-0
Formula:C₈H₁₂Br₂Pd
MDL Number:MFCD00799635