Order Entry
Czech Republic
ContactUsLinkComponent
Lead in isooctane standards, complete set for method ASTM D5059 Part C
Lead in isooctane standards, complete set for method ASTM D5059 Part C
Catalog # 45337.KS
CAS Number:  
Lead in isooctane standards, complete set for method ASTM D5059 Part C
Catalog # 45337.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Lead in Isooctane Standards, Complete Set
  • MDL No.:
    MFCD00134050
  • UN No.:
    1262
  • Physical form:
    Liquid
  • Molecular formula:
    Matrix: Isooctane
  • Appearance:
    Colourless
  • Test method:
    ASTM D5059 Part C

Specifications

About this item

Molecular Formula: Matrix: Isooctane
Physical Form: Liquid
Appearance: Colourless
UN No.: UN1262
MDL No.: MFCD00134050