Poly(styrene-divinylbenzene), cross-linked with 2% DVB 200-400 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9003-70-7
:9052-95-3
Formula:[-CH₂CH(C₆H₅)-]x[-CH₂CH[C₆H₄(CHCH₂)]-]y
MDL Number:MFCD00146442
:234.342 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together