
Palladium 10% on activated carbon powder, eggshell, reduced
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
Storage Temperature:Ambient
UN:3178
ADR:4.1,III
:GHS02
Related Information
Frequently Bought Together