Order Entry
Czech Republic
ContactUsLinkComponent
Silicon(IV) 2-ethylhexanoate ≥75% in ethanol
Flammable
Health
Silicon(IV) 2-ethylhexanoate ≥75% in ethanol
Catalog # 44635.30
CAS Number:  
Flammable
Health
Silicon(IV) 2-ethylhexanoate ≥75% in ethanol
Catalog # 44635.30
CAS Number:  

Specifications

  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :600.91 g/mol
Storage Temperature:Ambient
UN:1170
ADR:3,II
 :GHS02
 :GHS08
CAS Number:67939-81-5
Formula:C₃₂H₆₀O₈Si
MDL Number:MFCD00269842