

3,3'-Diethylthiadicarbocyanine iodide [DiSC₂(5)] ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:514-73-8
:
Formula:C₂₃H₂₃N₂S₂I
MDL Number:MFCD00074829
:518.48 g/mol
Merck Index:14,3369
Melting Pt:248 °C (dec.)
Storage Temperature:Ambient
EINECS:208-186-7
ADR:6.1,II
:GHS06
:GHS07
Related Information
Frequently Bought Together