Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 0.35 micron, carboxylated
Catalog # 43717.4Y
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Bal.:15 mL
- Sekvence:2295
Specifications
About this item
:
:125000 - 250000 g/mol
Storage Temperature:Refrigerator
CAS Number:9003-53-6
Formula:C₈H₈
MDL Number:MFCD00084450