
Ruthenium(III) nitrosylacetate ≥99.99% (metals basis), Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:Ru(NO)(OOCCH₃)₃
MDL Number:MFCD02684507
:308.18 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together