Order Entry
Czech Republic
ContactUsLinkComponent
Poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Catalog # 43292.14
CAS Number:  
Poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Catalog # 43292.14
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :234.342 g/mol
Storage Temperature:Ambient
CAS Number:9003-70-7
 :9052-95-3
Formula:[-CH₂CH(C₆H₅)-]x[-CH₂CH[C₆H₄(CHCH₂)]-]y
MDL Number:MFCD00146442