Order Entry
Czech Republic
ContactUsLinkComponent
 
Silicon dioxide 30% in ethylene glycol, colloidal dispersion
Catalog # 43093.A1
CAS Number:  
Silicon dioxide 30% in ethylene glycol, colloidal dispersion
Catalog # 43093.A1
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :60.08 g/mol
Density:1.3
Storage Temperature:Ambient
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232