Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 0.75 micron
Catalog # 42742.4Y
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:15 mL
Specifications
About this item
:
:125000 - 250000 g/mol
Storage Temperature:Refrigerator
CAS Number:9003-53-6
Formula:C₈H₈
MDL Number:MFCD00084450