Order Entry
Czech Republic
ContactUsLinkComponent
Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 6.0 micron
Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 6.0 micron
Catalog # 42716.AA
CAS Number:  
Poly(styrene) 2.5% (w/w) dispersion in water, latex microsphere 6.0 micron
Catalog # 42716.AA
CAS Number:  

Specifications

  • Pk:
    5 mL

Specifications

About this item

 :
 :125000 - 250000 g/mol
Storage Temperature:Refrigerator
CAS Number:9003-53-6
Formula:C₈H₈
MDL Number:MFCD00084450