
Poly(styrene sulphonic acid), M.W. M.W. 75,000 30% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:28210-41-5
:
Formula:[-CH₂CH(C₆H₄SO₃H)-]n
MDL Number:MFCD00165973
Density:1.1
Storage Temperature:Ambient
UN:2586
ADR:8,III
:GHS05
Related Information
Frequently Bought Together