Chromate, ion chromatography standard solution, Specpure®, CrO₄²⁻ 1000 µg/ml
Dodavatel: Thermo Fisher Scientific
Certifikáty
O tomto produktu
Molecular Formula: K₂CrO₄ in H₂O
Physical Form: Liquid
Appearance: Yellow
Odour: Odourless
MDL No.: MFCD00011368
Specifikace
- MDL No.:MFCD00011368
- Physical form:Liquid, yellow
- Odour/scent:Odourless
- Molecular formula:K2CrO4 in H2O
Specifikace
Související informace
Frequently Bought Together