Order Entry
Czech Republic
ContactUsLinkComponent
Cerium(IV) solution 0.25 N in dilute nitric acid etchant solution
Oxidizing
Corrosive
Cerium(IV) solution 0.25 N in dilute nitric acid etchant solution
Catalog # 42110.K7
CAS Number:  
Oxidizing
Corrosive
Cerium(IV) solution 0.25 N in dilute nitric acid etchant solution
Catalog # 42110.K7
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

About this item

 :
Storage Temperature:Ambient
UN:3098
ADR:5.1,III
 :GHS03
 :GHS05
MDL Number:MFCD01866052