
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64536-78-3
:
Formula:C₃₁H₅₀F₆IrNP₂
MDL Number:MFCD00075097
Melting Pt:175 °C (dec.)
:804.90 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together