ContactUsLinkComponent
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire, twilled weave 400 mesh woven from 0.028mm (0.0011in) Type 304
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire, twilled weave 400 mesh woven from 0.028mm (0.0011in) Type 304
Frequently Bought Together