Silicon(IV) nitride, MgO binder, sputtering target ≥99.9% (metals basis), Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41090.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Bal.:1
- Sekvence:0001
Specifications
About this item
:
Formula:Si₃N₄
MDL Number:MFCD00011230
:140.28 g/mol
Storage Temperature:Ambient
EINECS:234-796-8