Order Entry
Czech Republic
ContactUsLinkComponent
Silicon(IV) nitride, MgO binder, sputtering target ≥99.9% (metals basis), Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Silicon(IV) nitride, MgO binder, sputtering target ≥99.9% (metals basis), Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41088.KS
CAS Number:  
Silicon(IV) nitride, MgO binder, sputtering target ≥99.9% (metals basis), Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41088.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Formula:Si₃N₄
MDL Number:MFCD00011230
 :140.28 g/mol
Storage Temperature:Ambient
EINECS:234-796-8